missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ ABT 089 Dihydrochloride

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
ABT 089 dihydrochloride is a high affinity and selective α4β2 partial agonist (Ki = 16 nM). Exhibits >1000-fold and >10,000-fold selectivity for α4β2 over α1β1γ1 and α7 receptors respectively. Neuroprotective against glutamate-induced toxicity in rat cortical neurons in vitro. Enhances cognitive performance in vivo. Orally bioavailable and brain penetrant.
Specifications
Specifications
| Chemical Name or Material | ABT 089 dihydrochloride |
| CAS | 161416-61-1 |
| Synonym | 2-Methyl-3-[(2S)-pyrrolidinylmethoxy]pyridine dihydrochloride |
| Quantity | 10 mg |
| Solubility | Soluble to 100mM in water and to 20mM in DMSO |
| Molecular Formula | C11H16N2O·2HCl |
| Formula Weight | 265.18 |
| Physical Form | Solid |
| Product Type | Bioactive Small Molecule |
| Purity | >98% |
Research Use Only. Not For Human or Veterinary Use
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction