missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(4-Fluorobenzoyl)-propionic acid, Lipomed
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Description
3-(4-Fluorobenzoyl)-propionic acid
Specifications
Specifications
| Chemical Name or Material | 4-(4-Fluorophenyl)-4-oxobutanoic Acid |
| CAS | 366-77-8 |
| Analyte or Component Names | 4-(4-Fluorophenyl)-4-oxobutanoic Acid |
| Shipping Condition | Room Temperature |
| Molecular Formula | C10 H9 F O3 |
| Synonym | Benzenebutanoic acid, 4-fluoro-γ-oxo- (9 CI, ACI), 4-Fluoro-γ-oxobenzenebutanoic acid (ACI), Propionic acid, 3-(p-fluorobenzoyl)- (7 CI, 8 CI), 3-(4-Fluorobenzoyl)propanoic acid, 3-(4-Fluorobenzoyl)propionic acid, 3-(p-Fluorobenzoyl)propionic acid, 4-(4-Fluorophenyl)-4-oxobutanoic acid, 4-(4-Fluorophenyl)-4-oxobutyric acid, 4-Fluorobenzoylpropionic acid, 4-Oxo-4-(p-fluorophenyl)butyric acid, Ba 2833, NSC 408180, β-(4-Fluorobenzoyl)propionic acid, β-(p-Fluorobenzoyl)propionic acid, γ-(4-Fluorophenyl)-γ-oxobutanoic acid |
| InChI Formula | InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H |
| SMILES | OC(=O)CCC(=O)c1ccc(F)cc1 |
| IUPAC Name | 4-(4-fluorophenyl)-4-oxobutanoic acid |
| Industry Type | Pharmaceutical |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction