missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Toluidine-d7, TRC
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | 2-Toluidine-d7 |
| CAS | 68408-22-0 |
| Molecular Formula | C7 D7 H2 N |
| Quantity | 10 mg |
| Synonym | o-Toluidine-d7, 1-Amino-2-methylbenzene-d7, 2-Amino-1-methylbenzene-d7, 2-Aminotoluene-d7, 2-Methyl-1-aminobenzene-d7, 2-Methylaniline-d7, 2-Methylbenzenamine-d7, 2-Methylphenylamine-d7, 2-Tolylamine-d7, NSC 15348-d7, o-Aminotoluene-d7, o-Methylaniline-d7, o-Methylbenzenamine-d7 |
| InChI Formula | InChI=1S/C7H9N/c1-6-4-2-3-5-7(6)8/h2-5H,8H2,1H3/i1D3,2D,3D,4D,5D |
| SMILES | [2H]c1c([2H])c([2H])c(c(N)c1[2H])C([2H])([2H])[2H] |
| IUPAC Name | 2,3,4,5-tetradeuterio-6-(trideuteriomethyl)aniline |
| Molecular Weight (g/mol) | 114.196 |
| Formula Weight | 114.1174 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction