missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Oxo-3-hydroxy-LSD, Lipomed
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Description
2-Oxo-3-hydroxy-LSD
Specifications
Specifications
| Chemical Name or Material | 2-Oxo-3-hydroxy-LSD |
| CAS | 111295-09-1 |
| Analyte or Component Names | 2-Oxo-3-hydroxy-LSD |
| Shipping Condition | Room Temperature |
| Molecular Formula | C20 H25 N3 O3 |
| Synonym | 2-Oxo-3-hydroxy-LSD, 2,3-Dihydro-3-hydroxy-2-oxo Lysergide |
| InChI Formula | InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H |
| SMILES | CCN(CC)C(=O)[C@H]1 CN(C)[C@@H]2 CC3(O)C(=O)Nc4cccc(C2=C1)c34 |
| IUPAC Name | (6 aR,9 R)-N,N-diethyl-5 A-hydroxy-7-methyl-5-oxo-6,6 A,8,9-tetrahydro-4 H-indolo[4,3-fg]quinoline-9-carboxamide |
| Industry Type | Pharmaceutical |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction