missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3,5-Trimethylpyrazine-d10, TRC
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
Specifications
Specifications
| Chemical Name or Material | 2,3,5-Trimethylpyrazine-d10 |
| Molecular Formula | C7 D10 N2 |
| Quantity | 10 mg |
| InChI Formula | InChI=1S/C7H10N2/c1-5-4-8-6(2)7(3)9-5/h4H,1-3H3/i1D3,2D3,3D3,4D |
| SMILES | [2H]c1nc(c(nc1C([2H])([2H])[2H])C([2H])([2H])[2H])C([2H])([2H])[2H] |
| IUPAC Name | 2-deuterio-3,5,6-tris(trideuteriomethyl)pyrazine |
| Molecular Weight (g/mol) | 132.229 |
| Formula Weight | 132.147 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction