missing translation for 'onlineSavingsMsg'
Learn More
Learn More
17Alpha-Methyltestosterone, Lipomed
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Description
17Alpha-Methyltestosterone
Specifications
Specifications
| Chemical Name or Material | 17-alpha-Methyltestosterone |
| CAS | 58-18-4 |
| Analyte or Component Names | 17-alpha-Methyltestosterone |
| Shipping Condition | Room Temperature |
| Molecular Formula | C20 H30 O2 |
| Synonym | Methyltestosterone, 17 alpha-Methyltestosterone, 17-Hydroxy-17-methyl-3-keto-androstene-4, 17-Methyltestosteron, 17-Methyltestosterone, Androst-4-ene-17 alpha-methyl-17 beta-ol-3-one, Androst-4-ene-17 alpha-methyl-17 beta-ol-3-one, Androst-4-ene-17 alpha-methyl-17 beta-ol-3-one, Androst-4-ene-17 alpha-methyl-17 beta-ol-3-one, Androst-4-ene-17 alpha-methyl-17 beta-ol-3-one, Androst-4-ene-17 alpha-methyl-17 beta-ol-3-one, Androst-4-ene-17 alpha-methyl-17 beta-ol-3-one, Androst-4-ene-17 alpha-methyl-17 beta-ol-3-one, Androst-4-ene-17 alpha-methyl-17 beta-ol-3-one, Androst-4-ene-17 alpha-methyl-17 beta-ol-3-one, Androst-4-ene-17 alpha-methyl-17 beta-ol-3-one, Androst-4-ene-17 alpha-methyl-17 beta-ol-3-one, Androst-4-ene-17 alpha-methyl-17 beta-ol-3-one, Androst-4-ene-17 alpha-methyl-17 beta-ol-3-one, Androst-4-ene-17 alpha-methyl-17 beta-ol-3-one, Androst-4-ene-17 alpha-methyl-17 beta-ol-3-one, Androst-4-ene-17 alpha-methyl-17 beta-ol-3-one, Androst-4-ene-17 alpha-methyl-17 beta-ol-3-one, Androst-4-ene-17 alpha-methyl-17 beta-ol-3-one, Androst-4-ene-17 alpha-methyl-17 beta-ol-3-one, Androst-4-ene-17 alpha-methyl-17 beta-ol-3-one, Androst-4-ene-17 alpha-methyl-17 beta-ol-3-one, Androst-4-ene-17 alpha-methyl-17 beta-ol-3-one, Androst-4-ene-17 alpha-methyl-17 beta-ol-3-one, Androst-4-ene-17 alpha-methyl-17 beta-ol-3-one, Androst-4-ene-17 alpha-methyl-17 beta-ol-3-one, Androst-4-ene-17 alpha-methyl-17 beta-ol-3-one, Androst-4-ene-17 alpha-methyl-17 beta-ol-3-one, Androst-4-ene-17 alpha-methyl-17 beta-ol-3-one, Androst-4-ene-17 alpha-methyl-17 beta-ol-3-one, Androst-4-ene-17 alpha-methyl-17 beta-ol-3-one, Androst-4-ene-17 alpha-methyl-17 beta-ol-3-one, Androst-4-ene-17 alpha-methyl-17 beta-ol-3-one, Androst-4-ene-17 alpha-methyl-17 beta-ol-3-one, Androst-4-ene-17 alpha-methyl-17 beta-ol-3-one, Androst-4-ene-17 alpha-methyl-17 beta-ol-3-one, Androst-4-ene-17 alpha-methyl-17 beta-ol-3-one, Androst-4-ene-17 alpha-methyl-17 beta-ol-3-one, Androst-4-ene-17 alpha-methyl-17 beta-ol-3-one, Androst-4-ene-17 alpha-methyl-17 beta-ol-3-one, Androst-4-ene-17 alpha-methyl-17 beta-ol-3-one, Androst-4-ene-17 alpha-methyl-17 beta-ol-3-one, Androst-4-ene-17 alpha-methyl-17 beta-ol-3-one, Androst-4-ene-17 alpha-methyl-17 beta-ol-3-one, Androst-4-ene-17 alpha-methyl-17 beta-ol-3-one, Androst-4-ene-17 alpha-methyl-17 beta-ol-3-one, Androst-4-ene-17 alpha-methyl-17 beta-ol-3-one, Androst-4-ene-17 alpha-methyl-17 beta-ol-3-one, Androst-4-ene-17 alpha-methyl-17 beta-ol-3-one, Androst-4-ene-17 alpha-methyl-17 beta-ol-3-one, Androst-4-ene-17 alpha-methyl-17 beta-ol-3-one, Androst-4-ene-17 alpha-methyl-17 beta-ol-3-one, Androst-4-ene-17 alpha-methyl-17 beta-ol-3-one, Androst-4-ene-17 alpha-methyl-17 beta-ol-3-one, Androst-4-ene-17 alpha-methyl-17 beta-ol-3-one, Androst-4-ene-17 alpha-methyl-17 beta-ol-3-one |
| InChI Formula | InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H |
| SMILES | C[C@]1(O)CC[C@H]2 [C@@H]3 CCC4=CC(=O)CC[C@]4(C)[C@H]3 CC[C@]12 C |
| IUPAC Name | (8 R,9 S,10 R,13 S,14 S,17 S)-17-hydroxy-10,13,17-trimethyl-2,6,7,8,9,11,12,14,15,16-decahydro-1 H-cyclopenta[a]phenanthren-3-one |
| Industry Type | Pharmaceutical |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction