missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Xylenesulfonic Acid Sodium Salt, TRC
Click to view available options
Quantity:
10 g
Unit Size:
10g
Specifications
Specifications
| Chemical Name or Material | Xylenesulfonic Acid Sodium Salt |
| CAS | 1300-72-7 |
| Molecular Formula | C8H9NaO3S |
| Quantity | 10 g |
| Synonym | Dimethylbenzenesulfonic Acid Sodium Salt, Conco SXS, Cyclophil SXS 30, Eltesol SX 30, Eltesol SX 93, FAC 2, Kemmat SN 18, Naxonate, Naxonate 4L, Naxonate G, Naxonate SX, Richonate SXS, SXS 40, Sodium Dimethylbenzenesulfonate, Sodium Xylenesulfate, Sodium Xylenesulfonate, Stepanate SXS, Stepanate X, Ultrawet 40SX, Vanwet SXS 40, Witconate SXS |
| InChI Formula | InChI=1S/C8H10O3S.Na/c1-6-3-4-8(5-7(6)2)12(9,10)11;/h3-5H,1-2H3,(H,9,10,11);/q;+1/p-1 |
| SMILES | [Na+].Cc1ccc(cc1C)S(=O)(=O)[O-] |
| Molecular Weight (g/mol) | 208.21 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction