missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4-Dimethylaniline (2,4-Xylidine), TRC
Click to view available options
Quantity:
10 g
Unit Size:
10g
Specifications
Specifications
| Chemical Name or Material | 2,4-Dimethylaniline |
| CAS | 95-68-1 |
| Molecular Formula | C8 H11 N |
| Quantity | 10 g |
| Synonym | 2,4-Xylidine (8CI), 2,4-Dimethylbenzenamine, 1-Amino-2,4-dimethylbenzene, 2,4-Dimethylaniline, 2,4-Dimethylphenylamine, 2,4-Xylylamine, 4-Amino-1,3-dimethylbenzene, 4-Amino-1,3-xylene, NSC 7640, m-Xylidine |
| InChI Formula | InChI=1S/C8H11N/c1-6-3-4-8(9)7(2)5-6/h3-5H,9H2,1-2H3 |
| SMILES | Cc1ccc(N)c(C)c1 |
| IUPAC Name | 2,4-dimethylaniline |
| Molecular Weight (g/mol) | 121.18 |
| Formula Weight | 121.0891 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction