missing translation for 'onlineSavingsMsg'
Learn More

(10E)-10,11-Didehydro-11-deoxy-6-O-methylerythromycin, TRC

Product Code. 30134871
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30134871 50 mg 50mg
17903905 10 mg 10mg
17940835 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134871 Supplier Toronto Research Chemicals Supplier No. TRCD29753050MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 144604-03-5
Molecular Formula C38 H67 N O12
Molecular Weight (g/mol) 729.94
Synonym (10E)-10,11-Didehydro-11-deoxy-6-O-methylerythromycin A, Oxacyclotetradecane, erythromycin deriv.
IUPAC Name (3R,4S,5S,6R,7R,9R,11E,13S,14R)-6-[(2S,3R,4S,6R)-4-(dimethylamino)-3-hydroxy-6-methyloxan-2-yl]oxy-14-ethyl-13-hydroxy-4-[(2R,4R,5S,6S)-5-hydroxy-4-methoxy-4,6-dimethyloxan-2-yl]oxy-7-methoxy-3,5,7,9,11,13-hexamethyl-1-oxacyclotetradec-11-ene-2,10-dione
SMILES CC[C@H]1OC(=O)[C@H](C)[C@@H](O[C@H]2C[C@@](C)(OC)[C@@H](O)[C@H](C)O2)[C@H](C)[C@@H](O[C@@H]3O[C@H](C)C[C@@H]([C@H]3O)N(C)C)[C@@](C)(C[C@@H](C)C(=O)\C(=C\[C@]1(C)O)\C)OC

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C38H67NO12/c1-15-27-36(8,44)17-20(2)29(40)21(3)18-38(10,46-14)33(51-35-30(41)26(39(11)12)16-22(4)47-35)23(5)31(24(6)34(43)49-27)50-28-19-37(9,45-13)32(42)25(7)48-28/h17,21-28,30-33,35,41-42,44H,15-16,18-19H2,1-14H3/b20-17+/t21-,22-,23+,24-,25+,26+,27-,28+,30-,31+,32+,33-,35+,36+,37-,38-/m1/s1
Formula Weight 729.4663
Chemical Name or Material (10E)-10,11-Didehydro-11-deoxy-6-O-methylerythromycin A
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.