missing translation for 'onlineSavingsMsg'
Learn More

10b-Hydroxy ∆4-Tibolone, TRC

Product Code. 30401593
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30401593 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30401593 Supplier Toronto Research Chemicals Supplier No. TRCH96427025MG

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 10beta-Hydroxy delta4-Tibolone
CAS 105186-35-4
Molecular Formula C21 H28 O3
Quantity 25 mg
Synonym 19-Norpregn-4-en-20-yn-3-one, 17,19-dihydroxy-7-methyl-, (7α,17α)-, (7α,17α)-17,19-Dihydroxy-7-methyl-19-norpregn-4-en-20-yn-3-one, 10β-Hydroxy-Δ4-tibolone, 10,17-Dihydroxy-7α-methyl-19-nor-10β,17α-pregn-4-en-20-yn-3-one
InChI Formula InChI=1S/C21H28O3/c1-4-20(23)9-7-16-18-13(2)11-14-12-15(22)5-10-21(14,24)17(18)6-8-19(16,20)3/h1,12-13,16-18,23-24H,5-11H2,2-3H3/t13-,16+,17+,18+,19+,20+,21-/m1/s1
SMILES C[C@@H]1CC2=CC(=O)CC[C@]2(O)[C@H]3CC[C@@]4(C)[C@@H](CC[C@@]4(O)C#C)[C@H]13
IUPAC Name (7R,8S,9S,10S,13S,14S,17R)-17-ethynyl-10,17-dihydroxy-7,13-dimethyl-2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthren-3-one
Molecular Weight (g/mol) 328.45
Formula Weight 328.2038
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.