missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethanol ≥ 96% (v/v), BAKER ANALYZED™ ACS, J.T. Baker™
Click to view available options
Quantity:
1 L
2.5 L
Specifications
Melting Point | -114°C |
Color | Max. 10 (APHA) |
Assay Percent Range | 95.1 to 96.9% (v/v) |
Quantity | 2.5 L |
Residue after Evaporation | max. 0.001% |
Titratable Acid | max. 0.0005 meq/g |
Titratable Base | max. 0.0002 meq/g |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'