missing translation for 'onlineSavingsMsg'
Learn More

Acetohydroxamic acid, 98+%

Product Code. 10671752 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10671752 5 g Glass bottle 5g
10299833 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10671752 Supplier Thermo Scientific Acros Supplier No. 170130050
From Chemicals to Chromatography
Use promo code 27991
View all promotions
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 546-88-3 | C2H5NO2 | 75.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 546-88-3
Molecular Formula C2H5NO2
Molecular Weight (g/mol) 75.06
MDL Number MFCD00009994
InChI Key RRUDCFGSUDOHDG-UHFFFAOYSA-N
Synonym acetohydroxamic acid, methylhydroxamic acid, acetylhydroxamic acid, acetic acid, oxime, acetohydroximic acid, lithostat, n-acetylhydroxylamine, acetamide, n-hydroxy, acethydroxamsaure, acethydroxamsaeure
PubChem CID 1990
ChEBI CHEBI:49029
IUPAC Name N-hydroxyacetamide
SMILES CC(=O)NO

Especificaciones

CAS Min % 98.0
CAS Max % 100.0
Melting Point 86.0°C to 90.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Glass bottle
Linear Formula CH3C(=NOH)OH
Quantity 5 g
Beilstein 02, 187
Fieser 13,2
Merck Index 15, 62
Solubility Information Solubility in water: soluble
Formula Weight 75.06
Percent Purity 98+%
Physical Form Crystalline Solid
Chemical Name or Material Acetohydroxamic acid
Mostrar más Mostrar menos
compliance-icons
Nombre del producto
  • Acetohydroxamic acid
Palabra de advertencia
  • Danger
Clasificación
  • Reproductive toxicity Category 1B
Indicaciones de peligro
  • H360D-May damage the unborn child.
Consejos de prudencia
  • P201-Obtain special instructions before use.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.