Learn More
Thermo Scientific Maybridge Methyl tetrahydrothiopyran-4-carboxylate, 1g, 97%
Brand: Thermo Scientific Maybridge CC61922DA
Methyl tetrahydrothiopyran-4-carboxylate, 1g, 97%
Code : 31
Additional Details : CAS Number : 128094-82-6 Weight : 0.05100kg Transport : UN number : 2810 Chem class : 6.1 Pack group : III