Learn More
Thermo Scientific Maybridge 6-Amino-benzoxazole, 5g, 95%
Brand: Thermo Scientific Maybridge MO07249DE
6-Amino-benzoxazole, 5g, 95%
Code : 31
Additional Details : CAS Number : 177492-52-3 Weight : 0.05500kg Transport : UN number : 2811 Chem class : 6.1 Pack group : III