Learn More
Thermo Scientific Maybridge 2-Fluoro-3-iodopyridine, 10g, 97%
Brand: Thermo Scientific Maybridge MO00359EA
2-Fluoro-3-iodopyridine, 10g, 97%
Code : 31
Additional Details : CAS Number : 113975-22-7 Weight : 0.06000kg Transport : UN number : 2811 Chem class : 6.1 Pack group : III