Learn More
Thermo Scientific™ LanthaScreen™ Tb-anti-p53 [AcLys382] Antibody
For use in combination with the BacMam p53 [AcLys382] Cellular Assay (A12901) to evaluate inhibitors in medium to high throughput screening applications.
Brand: Thermo Scientific™ PV6110
This item is not returnable.
View return policy
Description
- The LanthaScreen™ antibody detects the acetylation of p53 at Lys382
- For use in combination with the BacMam p53 [AcLys382] Cellular Assay (A12901)
- Evaluates inhibitors in medium to high throughput screening applications
Specifications
| Tb (Terbium)/Lanthanide Chelates | |
| Terbium-labeled Anti-p53 [AcLys382] antibody in HEPES buffered saline. Store at -20°C. | |
| 340⁄490, 520 | |
| Acetylated | |
| LanthaScreen |
| 1 mg | |
| Frozen | |
| Microplate Reader | |
| p53 |