Learn More
Thermo Scientific Maybridge 3-Methyl-1,2-dihydropyridin-2-one, 5g, 97%
Brand: Thermo Scientific Maybridge CD09405DE
3-Methyl-1,2-dihydropyridin-2-one, 5g, 97%
Code : 31
Additional Details : CAS Number : 1003-56-1 Weight : 0.05500kg
Brand: Thermo Scientific Maybridge CD09405DE
3-Methyl-1,2-dihydropyridin-2-one, 5g, 97%
Code : 31
Additional Details : CAS Number : 1003-56-1 Weight : 0.05500kg