missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Maybridge Ethyl 1,4-dibenzylpiperazine-2-carboxylate, 1g, 97%

Brand:  Thermo Scientific Maybridge KM07907DA

Ethyl 1,4-dibenzylpiperazine-2-carboxylate, 1g, 97%

Code : 31

Additional Details : CAS Number : 72351-59-8 Weight : 0.05100kg

Product Code. 10304582

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.