Learn More
Thermo Scientific Maybridge Furan-2-sulfonyl chloride, 1g, 97%
Brand: Thermo Scientific Maybridge CC02003DA
Furan-2-sulfonyl chloride, 1g, 97%
Code : 31
Additional Details : CAS Number : 52665-48-2 Weight : 0.05100kg
Brand: Thermo Scientific Maybridge CC02003DA
Furan-2-sulfonyl chloride, 1g, 97%
Code : 31
Additional Details : CAS Number : 52665-48-2 Weight : 0.05100kg