missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TPH2 Recombinant Rabbit Monoclonal Antibody (PSH04-89)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA556513
This item is not returnable.
View return policy
Description
TPH2 Recombinant Monoclonal Antibody for IHC (F), IHC (P), IP, Western Blot
Tryptophan hydroxylase (TPH) is a member of the pterin-dependent aromatic acid hydroxylase family. It catalyzes the first and rate limiting step in the biosynthesis of serotonin, an important hormone and neurotransmitter. TPH is expressed predominantly in the brain stem. Mutations in this gene may be associated with psychiatric diseases such as bipolar affective disorder and major depression.
Specifications
| TPH2 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 0.1% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q8CGU9, Q8CGV2 | |
| Tph2 | |
| Recombinant protein with Human THP2 aa 1-490. | |
| 100 μL | |
| Primary | |
| Mouse, Rat | |
| Antibody | |
| IgG |
| Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| PSH04-89 | |
| Unconjugated | |
| Tph2 | |
| ADHD7; AU043594; MGC119994; Neuronal tryptophan hydroxylase; NTPH; Tph2; TPRH; TRPH; Tryptophan 5-hydroxylase 2; tryptophan 5-monooxygenase 2; tryptophan hydroxylase 2 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 216343, 317675 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction