missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ SODD/BAG4 Antibody (SR2489), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-27764
This item is not returnable.
View return policy
Description
SODD/BAG4 Monoclonal antibody specifically detects SODD/BAG4 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunocytochemistry/ Immunofluorescence
Specifications
| SODD/BAG4 | |
| Monoclonal | |
| Unconjugated | |
| 50 mM Tris-Glycine (pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Western Blot, Immunocytochemistry/Immunofluorescence | |
| SR2489 | |
| Western Blot 1:500-1:1000, Immunohistochemistry 1:50-1:100, Immunocytochemistry/ Immunofluorescence 1:50-1:200 | |
| BAG-4, BAG-family molecular chaperone regulator-4, Bcl-2-associated athanogene 4, BCL2-associated athanogene 4, Silencer of death domains, SODDBAG family molecular chaperone regulator 4 | |
| A synthetic peptide of human SODD/BAG4 (Uniprot #: O95429) | |
| 100 μL | |
| Apoptosis, Cancer, Mitochondrial Mediated Pathway, Tumor Suppressors | |
| 9530 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction