missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLC35A2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5148355
This item is not returnable.
View return policy
Description
SLC35A2 Polyclonal Antibody for IHC (P)
This gene encodes a member of the nucleotide-sugar transporter family. The encoded protein is a multi-pass membrane protein. It transports UDP-galactose from the cytosol into Golgi vesicles, where it serves as a glycosyl donor for the generation of glycans. Mutations in this gene cause congenital disorder of glycosylation type IIm (CDG2M). Multiple alternatively spliced transcript variants encoding distinct isoforms have been found for this gene.
Specifications
| SLC35A2 | |
| Polyclonal | |
| Unconjugated | |
| SLC35A2 | |
| AI327289; CDG2M; CDGX; Had1; Had-1; mUGT1; Sfc8; SLC35A2; solute carrier family (UDP-N-acetylglucosamine transporter), member 2; solute carrier family 35 (UDP-galactose transporter) member 2; solute carrier family 35 (UDP-galactose transporter), member 2; solute carrier family 35 (UDP-galactose transporter), member A2; solute carrier family 35 member A2; solute carrier family 35, member A2; UDP-galactose translocator; UDP-galactose transporter; UDP-Gal-Tr; UGALT; UGAT; UGT; UGT1; UGT2; UGTL | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 7355 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 100 μg/mL | |
| PBS with 0.03% ProClin 300; pH 7 | |
| P78381 | |
| SLC35A2 | |
| E. coli-derived Human SLC35A2 fragment. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction