missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PABPN1 Recombinant Rabbit Monoclonal Antibody (24GB365)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557610
This item is not returnable.
View return policy
Description
PABPN1 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
This gene encodes an abundant nuclear protein that binds with high affinity to nascent poly(A) tails. The protein is required for progressive and efficient polymerization of poly(A) tails at the 3' ends of eukaryotic transcripts and controls the size of the poly(A) tail to about 250 nt. At steady-state, this protein is localized in the nucleus whereas a different poly(A) binding protein is localized in the cytoplasm. This gene contains a GCG trinucleotide repeat at the 5' end of the coding region, and expansion of this repeat from the normal 6 copies to 8-13 copies leads to autosomal dominant oculopharyngeal muscular dystrophy (OPMD) disease. Related pseudogenes have been identified on chromosomes 19 and X. Read-through transcription also exists between this gene and the neighboring upstream BCL2-like 2 (BCL2L2) gene.
Specifications
| PABPN1 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| PABPN1 | |
| mPABII; Nuclear poly(A)-binding protein 1; OPMD; PAB2; PABII; PABP2; PABP-2; Pabp3; PABPN1; poly(A) binding protein 2; poly(A) binding protein II; poly(A) binding protein nuclear 1; poly(A) binding protein, nuclear 1; poly(A)-binding protein 2; poly(A)-binding protein II; polyA binding protein II; polyadenylate-binding nuclear protein 1; Polyadenylate-binding protein 2 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 116697, 54196, 8106 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB365 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q86U42, Q8CCS6 | |
| PABPN1 | |
| A synthesized peptide derived from human PABPN1 (aa 4-31). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction