missing translation for 'onlineSavingsMsg'
Learn More
Learn More
P2Y1 Recombinant Rabbit Monoclonal Antibody (HL2702)

P2Y1 Recombinant Rabbit Monoclonal Antibody (HL2702)
Brand: Invitrogen MA556822
This item is not returnable.
View return policy
Description
P2Y1 Recombinant Monoclonal Antibody for Western Blot
Specifications
| P2Y1 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| P47900 | |
| P2ry1 | |
| Synthetic peptide encompassing a sequence within the Intracellular domain of human P2Y1. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Western Blot | |
| HL2702 | |
| Unconjugated | |
| P2ry1 | |
| ADP receptor; ATP receptor; P2 purinoceptor subtype Y1; P2 purinoreceptor subclass 2Y; P2RY1; P2y; P2Y ATP receptor 1; P2Y purinoceptor 1; P2Y1; P2Y1 receptor; p2y1r; p2y-1-r; platelet ADP receptor; Purinergic receptor; purinergic receptor P2Y, G-protein coupled 1; purinergic receptor P2Y, G-protein coupled, 1; purinergic receptor P2Y1; Purinoceptor p2y1; Purinoceptor p2yr1 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 5028 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction