missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RhoGAP Antibody (OTI9H2), Novus Biologicals Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-44958-100ul
This item is not returnable.
View return policy
Description
RhoGAP Monoclonal antibody specifically detects RhoGAP in Human,Mouse,Rat samples. It is validated for Western Blot
Specifications
| RhoGAP | |
| Monoclonal | |
| Unconjugated | |
| PBS (pH 7.3) 1% BSA, 50% glycerol | |
| Mouse | |
| >95% | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG2b |
| Western Blot | |
| OTI9H2 | |
| Western Blot 1:500-2000 | |
| growth factor independent 2, p100 RasGAP-associated p105 protein, p105 RhoGAP, p190-BGFI2, p190BRhoGAP, Rho GTPase activating protein 5, rho GTPase-activating protein 5, RHOGAP5, Rho-type GTPase-activating protein 5 | |
| Human recombinant protein fragment corresponding to amino acids 948-1261 of human ARHGAP5 (NP_001164) produced in E.coli. | |
| 100 μL | |
| Cell Biology, GPCR, Signal Transduction | |
| 394 | |
| Store at -20 degrees C. Avoid freeze/thaw cycles. | |
| Purified |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu