missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MCD Recombinant Rabbit Monoclonal Antibody (HL2738)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA556785
This item is not returnable.
View return policy
Description
MCD Recombinant Monoclonal Antibody for Western Blot, IHC (P)
MCD catalyzes the breakdown of malonyl-CoA to acetyl-CoA and carbon dioxide. Malonyl-CoA is an intermediate in fatty acid biosynthesis, and also inhibits the transport of fatty acyl CoAs into mitochondria. Consequently, the encoded protein acts to increase the rate of fatty acid oxidation. It is found in mitochondria, peroxisomes, and the cytoplasm. Mutations in this gene result in malonyl-CoA decarboyxlase deficiency.
Specifications
| MCD | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| O95822, Q920F5, Q99J39 | |
| MLYCD | |
| Recombinant fragment of human MCD. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot | |
| HL2738 | |
| Unconjugated | |
| MLYCD | |
| AI324784; malonyl coenzyme A decarboxylase; malonyl-CoA decarboxylase; malonyl-CoA decarboxylase, mitochondrial; MCD; MLYCD | |
| Rabbit | |
| Protein A | |
| RUO | |
| 23417, 56690, 85239 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction