missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Invitrogen™ MAS1L Recombinant Rabbit Monoclonal Antibody (24GB140)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557065
Denne vare kan ikke returneres.
Se returpolicy
Beskrivelse
MAS1L Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
The proto-oncogene MAS1 is a G protein-coupled receptor located on the plasma membrane. In transfected NIH/3T3 cells, MAS1 has a weak focus-inducing activity. MAS1 is an antagonist of the AT1 receptor (Angiotensin II type 1 receptor), inhibiting the actions of Angiotensin II. MAS1L (MAS1-like), also known as MAS-L or MRG (Mas-related G-protein coupled receptor MRG), is a 378 amino acid multi-pass membrane protein that belongs to the G-protein coupled receptor 1 (GPCR) family. MAS1 related proteins are thought to regulate nociceptor function and development, including the sensation or modulation of pain.
Tekniske data
| MAS1L | |
| Recombinant Monoclonal | |
| Unconjugated | |
| MAS1L | |
| DAAP-341G9.4; dJ994E9.2; MAS1 oncogene-like; MAS1 proto-oncogene like, G protein-coupled receptor; MAS1L; MAS1-like; MAS-L; MAS-R; mas-related G protein-coupled MRG; Mas-related G protein-coupled receptor b; MAS-related gene H; MAS-related GPR, member H; mas-related G-protein coupled receptor member H; Mas-related G-protein coupled receptor MRG; MAS-related G-protein coupled receptor, member H; MGC119987; Mgrb; MRG; Mrg and gpcr; Mrgh; MrgHGPR90; Mrgprh | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 116511, 404641 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB140 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P35410, Q7TN38 | |
| MAS1L | |
| A synthesized peptide derived from human MAS1L (aa 342-375). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion