missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HRASLS Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5148405
This item is not returnable.
View return policy
Description
HRASLS Polyclonal Antibody for IHC (P)
Specifications
| HRASLS | |
| Polyclonal | |
| Unconjugated | |
| HRASLS | |
| A-C1; HRAS like suppressor; HRAS-like suppressor 1; HRASLS; HRASLS1; H-REV107; H-REV107 protein-related protein; HRSL1; HSD28; phospholipase A; Phospholipase A and acyltransferase 1; Phospholipid-metabolizing enzyme A-C1; PLAAT1 | |
| Rabbit | |
| Antigen Affinity Chromatography, Protein A | |
| RUO | |
| 57110 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 100 μg/mL | |
| PBS with 0.03% ProClin 300; pH 7 | |
| Q9HDD0 | |
| HRASLS | |
| E. coli-derived Human HRASLS fragment. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction