Learn More
FGFR2 Antibody (2E9), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP3-26583-100ul
 View more versions of this product
Product Code. 30043495
  • £406.00 / 100µL
Description
FGFR2 Monoclonal antibody specifically detects FGFR2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY
Specifications
Specifications
FGFR2
Monoclonal
Unconjugated
PBS, pH 7.4, 150mM NaCl and 50% glycerol
Rabbit
Affinity purified
RUO
Primary
Human, Mouse, Rat
Purified
Western Blot, ELISA
2E9
Western Blot 1:500-1:5000, ELISA
BBDS, BEK, BFR-1, CD332, CD332 antigen, CEK3, CFD1, craniofacial dysostosis 1, EC 2.7.10, EC 2.7.10.1, ECT1, fibroblast growth factor receptor 2, FLJ98662, Jackson-Weiss syndrome, JWS, Keratinocyte growth factor receptorreceptor like 14, KGFR, K-sam, KSAM, soluble FGFR4 variant 4, TK14, TK25
A synthesized peptide derived from Human FGFR2 [UniProt P21802]
100 μL
Angiogenesis, Protein Kinase
2263
Store at -20 to -70C. Avoid freeze-thaw cycles.