missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ eIF3e Recombinant Rabbit Monoclonal Antibody (23GB5915), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA552948
This item is not returnable.
View return policy
Description
Component of the eukaryotic translation initiation factor 3 (eIF-3) complex, which is required for several steps in the initiation of protein synthesis. The eIF-3 complex associates with the 40S ribosome and facilitates the recruitment of eIF-1, eIF-1A, eIF-2:GTP:methionyl-tRNAi and eIF-5 to form the 43S preinitiation complex (43S PIC). The eIF-3 complex stimulates mRNA recruitment to the 43S PIC and scanning of the mRNA for AUG recognition. The eIF-3 complex is also required for disassembly and recycling of post termination ribosomal complexes and subsequently prevents premature joining of the 40S and 60S ribosomal subunits prior to initiation. Required for nonsense-mediated mRNA decay (NMD); may act in conjunction with UPF2 to divert mRNAs from translation to the NMD pathway. May interact with MCM7 and EPAS1 and regulate the proteasome-mediated degradation of these proteins.
Specifications
| eIF3e | |
| Recombinant Monoclonal | |
| Unconjugated | |
| EIF3E | |
| 48kDa; eIF-3 p48; eIF3e; eIF3-p46; EIF3-P48; EIF3S6; eukaryotic translation initiation factor 3 subunit 6; Eukaryotic translation initiation factor 3 subunit E; eukaryotic translation initiation factor 3 subunit E isoform 2 transcript; eukaryotic translation initiation factor 3 subunit E-like protein; eukaryotic translation initiation factor 3, subunit 6 (48kD); eukaryotic translation initiation factor 3, subunit 6 48kDa; eukaryotic translation initiation factor 3, subunit E; eukaryotic translation initiation factor subunit E-like protein; Int6; mammary tumor integration site 6; mammary tumor-associated protein INT6; mammary tumor-associated protein INT-6; MMTV integration site 6; murine mammary tumor integration site 6 (oncogene homolog); NEWGENE_1311521; Viral integration site protein INT-6; viral integration site protein INT-6 homolog | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 103692906, 16341, 3646 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB5915 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P60228, P60229, Q641X8 | |
| EIF3E | |
| A synthesized peptide derived from human eIF3e (210-350AA). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction