missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CNGA4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5145067
This item is not returnable.
View return policy
Description
Immunogen sequence: LLAELESSAL KIAYRIERLE WQTREWPMPE DLAEADDEGE PEEGTSKDEE GRAS Antigen sequence identity: Mouse 83%; Rat 87%.
Second messenger, cAMP, causes the opening of cation- selective cyclic nucleotide-gated (CNG) channels and depolarization of the neuron (olfactory sensory neurons, OSNs). CNGA4 is the modulatory subunit of this channel which is known to play a central role in the transduction of odorant signals and subsequent adaptation. By accelerating the calcium-mediated negative feedback in olfactory signaling it allows rapid adaptation to odor stimulation and extends its range of odor detection.
Specifications
| CNGA4 | |
| Polyclonal | |
| Unconjugated | |
| CNGA4 | |
| Cgn2; CNCA2; CNCalpha4, CNG5; cng channel; CNG channel alpha-4; CNG4; CNG-4; CNG5; CNGA4; CNGB2; cyclic nucleotide gated cation channel; cyclic nucleotide gated channel alpha 4; cyclic nucleotide gated channel beta 2; cyclic nucleotide-gated cation channel alpha 4; cyclic nucleotide-gated cation channel alpha-4; Cyclic nucleotide-gated channel alpha-4; cyclic nucleotide-gated olfactory channel subunit OCNC2; OCNC2; OCNCb; OCNCBETA; olfactory cyclic nucleotide-gated channel; rOCNC2 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 1262 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid | |
| IgG |
| Immunocytochemistry | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8IV77 | |
| CNGA4 | |
| Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 514-567. Recombinant protein control fragment (Product #RP-109765). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction