missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD42b Recombinant Mouse Monoclonal Antibody (13G2-R)

Mouse Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA556390
This item is not returnable.
View return policy
Description
CD42b Recombinant Monoclonal Antibody for IHC (P)
CD42b is a platelet activation marker involved in the process of coagulation as an aggregating factor. It interacts with extracellular matrix as well as with adhesion molecules. CD42b expression and activation seems to be regulated by VEGF and PDGF. CD42b composes together with GPIb beta, GPIX and GPV the GPIb-IX-V receptor complex critical in the process of platelet-rich thrombus formation by tethering the platelet to a thrombogenic surface. CD42b binds to von Willebrand factor (VWF) exposed at a site of vascular injury, as well as to thrombin, coagulation factors XI and XII, high molecular wight kininogen, TSP-1, integrin Mac-1 and P-selectin. The extracellular domain of CD42b by its interactions also contributes to metastasis. Further, CD42b antigen is expressed on platelets and on megakaryocytes in bone marrow. Several polymorphisms and mutations have been described in the CD42b, some of which are the cause of Bernard-Soulier syndromes and platelet-type von Willebrand disease.
Specifications
| CD42b | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 0.1% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| P07359 | |
| Gp1ba | |
| Recombinant protein within human CD42b aa 100-350/652. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin) | |
| 13G2-R | |
| Unconjugated | |
| Gp1ba | |
| Antigen CD42b-alpha; BDPLT1; BDPLT3; BSS; CD42a; CD42B; CD42b-alpha; DBPLT3; Glycocalicin; glycoprotein 1b, alpha polypeptide; glycoprotein Ib (platelet), alpha polypeptide; glycoprotein Ib platelet alpha subunit; glycoprotein Ib platelet subunit alpha; glycoprotein Ibalpha; GP1B; GP1BA; GPIB; GPIb alpha; GP-Ib alpha; GPIbA; GPIbalpha; GPIX; GP-IX; MGC34595; mutant platelet membrane glycoprotein Ib-alpha; platelet glycoprotein Ib alpha chain; platelet glycoprotein Ib alpha polypeptide; Platelet GPIX; platelet membrane glycoprotein 1b-alpha subunit; platelet membrane glycoprotein Ib-alpha; VWDP | |
| Mouse | |
| Protein A | |
| RUO | |
| 2811 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction