missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human FABP8/M-FABP Protein

Product Code. 18718228
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
This item is not returnable. View return policy

Product Code. 18718228

Brand: R&D Systems™ 9487CL100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

R&D Systems™ Recombinant Human FABP8/M-FABP Protein CF is a member of a large superfamily of lipid binding proteins that are expressed in a tissue specific manner.

The Recombinant Human FABP8/M-FABP Protein is derived from E. coli.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P02689
Characterization Bioactivity not tested
For Use With (Application) Flow Cytometry
Formulation Lyophilized from a 0.2μm filtered solution in Sodium Acetate.
Gene ID (Entrez) 5375
Molecular Weight (g/mol) 16kDa
Quantity 100 μg
Source E. coli-derived human FABP8/M-FABP protein Ser2-Val132,with a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10EU per 1μg of the protein by the LAL method.
Gene Alias MFABP, M-FABP, MP2, myelin P2 protein, P2, peripheral myelin protein 2FABP8, PMP2
Product Type Recombinant Protein
Conjugate Unconjugated
Species Human
Reconstitution Reconstitute at 1mg/mL in water.
Purity or Quality Grade >90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.