missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rat Endoglin/CD105 Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF6440
This item is not returnable.
View return policy
Description
Endoglin/CD105 Polyclonal specifically detects Endoglin/CD105 in Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunocytochemistry.
Specifications
| Endoglin/CD105 | |
| Polyclonal | |
| Western Blot 1 ug/mL, Immunohistochemistry 5-15 ug/mL, Immunoprecipitation 4 ug/mL, Immunocytochemistry 5-15 ug/mL | |
| Q6Q3E8 | |
| ENG | |
| Mouse myeloma cell line NS0-derived recombinant rat Endoglin/CD105 Met1-Gly578 Accession # Q6Q3E8 | |
| 100 μg | |
| Primary | |
| Detects rat Endoglin/CD105 in direct ELISAs and Western blots. In direct ELISAs, approximately 75% cross reactivity with recombinant mouse Endoglin is observed and approximately 50% cross reactivity with recombinant human Endoglin is observed. | |
| Rat | |
| IgG |
| Western Blot, Immunohistochemistry, Immunoprecipitation, Immunocytochemistry | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD105, CD105 antigen, endoglin, ENDOsler-Rendu-Weber syndrome 1, ENG, HHT1FLJ41744, ORW, ORW1 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 2022 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction