missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Escherichia T7 phage RNA polymerase Protein, CF
Click to view available options
:
100 μg
Description
Measured by its ability to produce 4.1 kB RNA per agarose gel electrophoresis from a defined linearized DNA plasmid template quantified using a fluorescent detection assay. The RNA yield is >2500 ng, as measured under the described conditions.
Specifications
Specifications
| Accession Number | P00573.2 |
| For Use With (Application) | Enzyme Activity |
| Formulation | Supplied as a 0.2 μm filtered solution in Sodium Phosphate, NaCl, TCEP and Glycerol. |
| Gene ID (Entrez) | 1261050 |
| Molecular Weight (g/mol) | MolecularWeight-observed: 89-98 kDa, under reducing conditions, MolecularWeight-theroretical: 100 kDa |
| Quantity | 100 μg |
| Source | E. coli-derived e. coli T7 phage RNA polymerase protein Asn2-Ala883 with a N-terminal Met and 6-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70 °C as supplied. 3 months, -20 to -70 °C under sterile conditions after opening. |
| Endotoxin Concentration | <0.10 EU per 1 μg of the protein by the LAL method. |
| Gene Alias | T7p07 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction