Learn More
Novus Biologicals™ Nse2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals™ NBP3-30136
Product Code. 30044780
  • £432.00 / 100µg
Description
Nse2 Polyclonal antibody specifically detects Nse2 in Human samples. It is validated for Immunoprecipitation
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Nse2
Polyclonal
Tris-citrate/phosphate buffer, pH 7 to 8
Rabbit
Affinity purified
RUO
286053
Store at 4°C. Do not freeze.
IgG
Immunoprecipitation
Unconjugated
C8orf36, E3 SUMO-protein ligase NSE2, EC 6.3.2, EC 6.3.2.-, FLJ32440, hMMS21, methyl methanesulfonate sensitivity gene 21, MMS21 homolog, MMS21chromosome 8 open reading frame 36, Non-SMC element 2 homolog, non-SMC element 2 homolog (MMS21, S. cerevisiae), non-SMC element 2, MMS21 homolog (S. cerevisiae), Non-structural maintenance of chromosomes element 2 homolog, NSE2
Between 125 and 175
100 μg
Primary
Human
Purified