missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ MED26 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-29345
This item is not returnable.
View return policy
Description
MED26 Polyclonal antibody specifically detects MED26 in Human samples. It is validated for Immunoprecipitation
Specifications
| MED26 | |
| Polyclonal | |
| Tris-citrate/phosphate buffer, pH 7 to 8 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 9441 | |
| Store at 4°C. Do not freeze. | |
| IgG |
| Immunoprecipitation | |
| Unconjugated | |
| Activator-recruited cofactor 70 kDa component, Cofactor required for Sp1 transcriptional activation subunit 7, cofactor required for Sp1 transcriptional activation, subunit 7 (70kD), cofactor required for Sp1 transcriptional activation, subunit 7, 70kDa, CRSP70ARC70, CRSP7CRSP complex subunit 7, mediator complex subunit 26Transcriptional coactivator CRSP70, mediator of RNA polymerase II transcription subunit 26 | |
| between 1 and 50 | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction