missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ DIAPH3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-29539
This item is not returnable.
View return policy
Description
DIAPH3 Polyclonal antibody specifically detects DIAPH3 in Human samples. It is validated for Immunoprecipitation,Western Blot
Specifications
| DIAPH3 | |
| Polyclonal | |
| Tris-citrate/phosphate pH 7 -8. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| DIAP3, diaphanous homolog 3 (Drosophila), diaphanous-related formin 3, DKFZp434C0931, DKFZp686A13178, homolog) 3, mDia2, protein diaphanous homolog 3 | |
| Between 1 and 50 | |
| 100 μg | |
| Cancer | |
| 81624 | |
| Store at 4°C. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction