missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ DAGLB Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-29738
This item is not returnable.
View return policy
Description
DAGLB Polyclonal antibody specifically detects DAGLB in Human samples. It is validated for Immunoprecipitation,Western Blot
Specifications
| DAGLB | |
| Polyclonal | |
| Tris-citrate/phosphate buffer, pH 7 to 8 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| beta, DAGLB, DAGLBETA, diacylglycerol lipase, diacylglycerol lipase beta, FLJ33624, FLJ33909, KCCR13L | |
| Between 622 and 672 | |
| 100 μg | |
| Cardiovascular Biology, Endocrinology, Signal Transduction | |
| 221955 | |
| Store at 4°C. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction