Learn More
COL4A3 Antibody (COL4A3/9395R), Novus Biologicals Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP3-45074
Product Code. 30448909
  • £450.00 / 100µg
Define Immunology Research on Your Terms With R&D Systems
Use promo code 24090
View all promotions
Description
COL4A3 Monoclonal antibody specifically detects COL4A3 in Human samples. It is validated for Immunohistochemistry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
COL4A3
Monoclonal
1 mg/mL
Immunohistochemistry : 1-2ug/mL
ATS2, ATS3, collagen alpha-3(IV) chain, collagen IV, alpha-3 polypeptide, collagen, type IV, alpha 3 (Goodpasture antigen), tumstatin
Recombinant fragment (around aa1400-1670) of human COL4A3 protein (Uniprot: P02462, P08572, P29400, P53420). Exact sequence is proprietary)
100 μg
Primary
Human
Antibody
IgG κ
Immunohistochemistry
COL4A3/9395R
Unconjugated
10 mM PBS
Rabbit
Protein A or G purified
RUO
1285
Store at -20 to -84°C. Avoid freeze-thaw cycles.
Purified