missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Oligodendrocyte Marker O4 Alexa Fluor™ 750-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB1326S-100UG
This item is not returnable.
View return policy
Description
Oligodendrocyte Marker O4 Monoclonal specifically detects Oligodendrocyte Marker O4 in Human, Mouse, Rat, Chicken samples. It is validated for Flow Cytometry.
Specifications
| Oligodendrocyte Marker O4 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat, Chicken | |
| IgM |
| Flow Cytometry | |
| O4 | |
| Alexa Fluor 750 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| Bovine brain corpus callosum white matter | |
| RUO | |
| Detects human, mouse, rat, and chicken Oligodendrocyte Marker O4. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction