missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse TLR11 Alexa Fluor 488-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems IC7640G-100UG
This item is not returnable.
View return policy
Description
TLR11 Monoclonal specifically detects TLR11 in Mouse samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
| TLR11 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
| Q6R5P0 | |
| TLR11 | |
| E. coli-derived recombinant mouse TLR11 Thr209-Gln325 Accession # Q6R5P0 | |
| RUO | |
| 442887 | |
| Mouse | |
| IgG2b |
| Flow Cytometry | |
| 786404 | |
| Alexa Fluor 488 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| Gm287, hgnc31753, Toll like receptor 11 | |
| Rat | |
| 100 μg | |
| Primary | |
| Detects mouse TLR11 in direct ELISAs. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction