missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FUT9 Antibody (2B6), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-42972
This item is not returnable.
View return policy
Description
FUT9 Monoclonal antibody specifically detects FUT9 in Human samples. It is validated for ELISA,Immunohistochemistry,Western Blot
Specifications
| FUT9 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.3, 50% glycerol | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 10690 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG1 |
| ELISA, Immunohistochemistry, Western Blot | |
| 2B6 | |
| Western Blot 1:500 - 1:2000, ELISA, Immunohistochemistry 1:20 - 1:200 | |
| alpha-(1,3)-fucosyltransferase, EC 2.4.1, EC 2.4.1.-, EC 2.4.1.65, Fucosyltransferase 9, fucosyltransferase 9 (alpha (1,3) fucosyltransferase), Fucosyltransferase IX, fucT-IX, Fuc-TIXFucT-IX, Galactoside 3-L-fucosyltransferase | |
| fucosyltransferase 9(alpha(1,3) fucosyltransferase) - (Uniprot# Q9Y231) | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction