missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DDB1 Antibody (3A0), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-42957
This item is not returnable.
View return policy
Description
DDB1 Monoclonal antibody specifically detects DDB1 in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot
Specifications
| DDB1 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.3, 50% glycerol | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| ELISA, Immunohistochemistry, Western Blot | |
| 3A0 | |
| Western Blot 1:500 - 1:2000, ELISA, Immunohistochemistry 1:200 - 1:800 | |
| damage-specific DNA binding protein 1 (127kD), damage-specific DNA binding protein 1, 127kDa, Damage-specific DNA-binding protein 1, DDB p127 subunit, DDBa, DNA damage-binding protein 1, DNA damage-binding protein a, HBV X-associated protein 1, UV-damaged DNA-binding factor, UV-damaged DNA-binding protein 1, UV-DDB 1, UV-DDB1, XAP1, XAP-1, Xeroderma pigmentosum group E-complementing protein, XPCe, XPE, XPE-BF, XPE-binding factor | |
| damage-specific DNA binding protein 1, 127kDa - (Uniprot# Q16531) | |
| 100 μg | |
| DNA Repair, Nucleotide Excision Repair | |
| 1642 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction