missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATPG Antibody (8H11), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-42931
This item is not returnable.
View return policy
Description
ATPG Monoclonal antibody specifically detects ATPG in Human samples. It is validated for ELISA,Immunohistochemistry,Western Blot
Specifications
| ATPG | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.3, 50% glycerol | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA, Immunohistochemistry, Western Blot | |
| 8H11 | |
| Western Blot 1:500 - 1:2000, ELISA, Immunohistochemistry 1:100 - 1:500 | |
| ATP synthase gamma chain, mitochondrial, ATP synthase, H+ transporting, mitochondrial F1 complex, gamma polypeptide 1, ATP5CATP synthase subunit gamma, mitochondrial, ATP5CL1mitochondrial ATP synthase, gamma subunit 1, F-ATPase gamma subunit | |
| ATP synthase, H+ transporting, mitochondrial F1 complex, gamma polypeptide 1 - (Uniprot# P36542) | |
| 100 μg | |
| Endocrinology, Neurodegeneration, Neuroscience, Signal Transduction | |
| 509 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction