missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATPB Antibody (5B3), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-43063
This item is not returnable.
View return policy
Description
ATPB Monoclonal antibody specifically detects ATPB in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot
Specifications
| ATPB | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.3, 50% glycerol | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| ELISA, Immunohistochemistry, Western Blot | |
| 5B3 | |
| Western Blot 1:5000 - 1:30000, ELISA, Immunohistochemistry 1:200 - 1:1000 | |
| ATP synthase subunit beta, mitochondrial, ATP synthase, H+ transporting, mitochondrial F1 complex, beta polypeptide, ATPMB, ATPSBmitochondrial ATP synthase beta subunit, EC 3.6.3, EC 3.6.3.14, MGC5231, mitochondrial ATP synthetase, beta subunit | |
| ATP synthase, H+ transporting, mitochondrial F1 complex, beta polypeptide - (Uniprot# P06576) | |
| 100 μg | |
| Endocrinology, Neurodegeneration, Neuroscience, Signal Transduction | |
| 506 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction