missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RNA polymerase I termination factor Antibody (7C4), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-43049
This item is not returnable.
View return policy
Description
RNA polymerase I termination factor Monoclonal antibody specifically detects RNA polymerase I termination factor in Human,Mouse samples. It is validated for ELISA,Western Blot
Specifications
| RNA polymerase I termination factor | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.3, 50% glycerol | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 7270 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG1 |
| ELISA, Western Blot | |
| 7C4 | |
| Western Blot 1:1000 - 1:10000, ELISA | |
| RNA polymerase I termination factor, transcription termination factor 1, Transcription termination factor I, transcription termination factor, RNA polymerase I, TTF-1, TTF-I | |
| transcription termination factor, RNA polymerase I - (Uniprot# Q15361) | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction