missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nicotinic Acetylcholine R alpha 7/CHRNA7 Antibody (4B8), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-42941
This item is not returnable.
View return policy
Description
Nicotinic Acetylcholine R alpha 7/CHRNA7 Monoclonal antibody specifically detects Nicotinic Acetylcholine R alpha 7/CHRNA7 in Human samples. It is validated for ELISA,Western Blot
Specifications
| Nicotinic Acetylcholine R alpha 7/CHRNA7 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.3, 50% glycerol | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA, Western Blot | |
| 4B8 | |
| Western Blot 1:500 - 1:2000, ELISA | |
| a7 nicotinic acetylcholine receptor, alpha 7 neuronal nicotinic acetylcholine receptor, alpha-7 nicotinic cholinergic receptor subunit, cholinergic receptor, nicotinic, alpha 7, cholinergic receptor, nicotinic, alpha polypeptide 7, CHRNA7-2, NACHRA7, neuronal acetylcholine receptor protein, alpha-7 chain, neuronal acetylcholine receptor subunit alpha-7 | |
| cholinergic receptor, nicotinic, alpha 7 - (Uniprot# P36544) | |
| 100 μg | |
| Signal Transduction | |
| 1139 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction