missing translation for 'onlineSavingsMsg'
Learn More
Learn More
INPP5F Antibody (OTI1D4), Novus Biologicals Biologicals™
Mouse Monoclonal Antibody
Marca: Novus Biologicals NBP3-44844
Les retours ne sont pas autorisés pour ce produit.
Consulta la politica di reso
Descrizione
INPP5F Monoclonal antibody specifically detects INPP5F in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot
Specifica
| INPP5F | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized powder ( 1X PBS, pH 7.3,) | |
| Mouse | |
| >95% | |
| RUO | |
| 22876 | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG1 |
| Immunohistochemistry, Western Blot | |
| OTI1D4 | |
| Western Blot 1:2000, Immunohistochemistry 1:500 | |
| EC 3.1.3, EC 3.1.3.-, EC 3.1.3.16, EC 3.1.3.36, FLJ13081, hSac2, Inositol polyphosphate 5-phosphatase F, inositol polyphosphate-5-phosphatase F, KIAA0966MGC59773, MGC131851, phosphatidylinositide phosphatase SAC2, Sac domain-containing inositol phosphatase 2, Sac domain-containing phosphoinositide 5-phosphatase 2, SAC2MSTPO47 | |
| Full length human recombinant protein of human INPP5F (NP_055752) produced in HEK293T cell. | |
| 100 μg | |
| Primary | |
| For reconstitution, we recommend adding 100uL distilled water to a final antibody concentration of about 1 mg/mL. To use this carrier-free antibody for conjugation experiment, we strongly recommend performing another round of desalting process. | |
| Store at -20 degrees C. Avoid freeze/thaw cycles. | |
| Lyophilized |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto