missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aquaporin-4 Antibody (AQP4/3323), Novus Biologicals Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-45054
This item is not returnable.
View return policy
Description
Aquaporin-4 Monoclonal antibody specifically detects Aquaporin-4 in Human samples. It is validated for Flow Cytometry,Immunohistochemistry,Western Blot
Specifications
| Aquaporin-4 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot : 2-4ug/mL, Flow Cytometry : 1-2ug/million cells, Immunohistochemistry : 1-2ug/mL | |
| aquaporin 4, MGC22454 | |
| Human recombinant Aquaporin-4 protein fragment (around aa200-323) (Uniprot: P55087). Exact sequence is proprietary. | |
| 100 μg | |
| Cancer, Endocrinology, Neurodegeneration, Neuroscience, Signal Transduction | |
| 361 | |
| Store at -20 to -84°C. Avoid freeze-thaw cycles. | |
| Purified |
| Flow Cytometry, Immunohistochemistry, Western Blot | |
| AQP4/3323 | |
| Unconjugated | |
| 10 mM PBS | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction